missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 96-Well Polypropylene UniPlate™ Microplates
Manufactured from polypropylene for superior chemical and thermal stability. Whatman™ 96-Well Polypropylene UniPlate™ Microplates withstand temperatures to -80°C for long-term storage.
Brand: Cytiva 7701-5200
189.06 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- One-piece molded collection and analysis plates
- Conforms to ANSI/SBS microplate standard
- Prevent optical cross talk in light emitting assays
- Untreated
- Ideal for filtrate collection, homogeneous assay techniques used in high-throughput screening (HTS)
Specifications
Standard | |
8 x 12 | |
Polypropylene | |
96 | |
Square, Round Bottom (U-well) |
UNIPLATE microplates | |
Natural | |
2 mL | |
Non-treated | |
For homogeneous assays techniques utilized in high-throughput screening |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction