Learn More
Cytiva Whatman™ Antibiotic Assay Discs
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Brand:  Cytiva 2017-013
 View more versions of this product
Product Code. 11350654
  • 259.00 € / Pack of 1000
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
  • For use with Kirby-Bauer method of antibiotic testing
  • Allows rational and selective chemotherapy
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Antibiotic Assay Discs
1000 Discs/Pk.
13 mm
Infectious Disease, Antibiotic Susceptibility Sensitivity
1000 Pack