Learn More
Cytiva Whatman™ AS 600 Series Filtration Manifold
Stainless steel filter funnel six-place manifold, for microbiology monitors and analytical funnels
Brand:  Cytiva 10498762
1820.98 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 11313125
  • 2040.00 € / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The stainless steel manifold for three or six filtration units is fitted with stainless steel units. The apparatus can be autoclaved and sterilized by dry heat at up to 180°C. Suitable only for vacuum operation. If flushing tubes are used, do not exceed 1.3 bar (300 mbar over-pressure).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Stainless Steel
9 mm I.D.
Microbiology monitors and analytical funnels
6 Stopcocks and lower parts for individual choice of filter units