missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Cellulose Nitrate Membrane - Grade NC45
Featuring very narrow pore size distribution and low levels of extractables.
Brand: Cytiva 10401126
This item is not returnable.
View return policy
Description
- Increased temperature stability
- Higher strength and flexibility
- Reduced shrinkage
- White plain filters
- Pore size: 0.45μm
Specifications
Autoclavable | |
Cellulose Nitrate | |
Non-sterile | |
NC45 | |
Plain | |
176°F | |
Membrane Filter | |
11 cm |
White | |
0.45 μm | |
Bulk bacterial removal (general), sediment analysis (water microbiology and analysis) | |
Hydrophilic | |
50 Pack | |
80°C | |
4.33 in. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger||Warning
Hazard Category
- Flammable solid Category 1
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose