missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desiccant Pack for 903 Protein Saver Cards, Whatman™
Click to view available options
Quantity:
100 g
Description
- For use with 903 Protein Saver cards
Specifications
Specifications
Physical Form | Solid |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction