missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Elute Micro
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
225.00 € - 705.00 €
Specifications
Content And Storage | Room Temperature |
---|---|
Description | Collect, process, transport and store colored biological samples, such as blood samples with Non-Indicating FTA Elute Micro Cards. |
Sample Type | Blood |
For Use With (Application) | Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection |
No. of Samples | 4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11938117
|
QIAGEN
WB120401 |
25 Pk. |
225.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11994416
|
QIAGEN
WB120410 |
100 Pk. |
705.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For collection, stabilization, processing, transport and archiving of biological samples. 4 sample areas per card.
- FTA Elute technology for convenient water based elution of nucleic acids.
- QIAcard FTA Elute Micro format with 4 spot area (spot volume 12 to 40 μL) to collect, stabilize, process, transport, and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Room Temperature | |
Blood | |
4 | |
Chaotropic | |
15 to 30 min |
Collect, process, transport and store colored biological samples, such as blood samples with Non-Indicating FTA Elute Micro Cards. | |
Multiplex PCR, Sequencing after PCR amplification, SNP analysis, STR analysis, HLA typing, Whole genome amplification, Quantitative PCR, Biobanking, Pharmacogenomics, Genotyping, Genetic identification, Transgenic detection | |
FTA Elute Micro Card | |
40 μL |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title