Learn More
Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper
17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
Brand:  Cytiva 3017-8793
 View more versions of this product
Product Code. 10622245
  • 97.80 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
  • Also widely used in protein and nucleic acid blotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
17 Chr cellulose chromatography roll is a 0.92 mm paper highly absorbent paper for preparative paper chromatography and electrophoresis.
0.92 mm
Sheet
8.66 in.
0.98 in.
190 mm/30 min.
2.5 x 22 cm (0.98 x 8.66 in.)
Protein and Nucleic Acid Blotting, Preparative Paper Chromatography and Electrophoresis
Grade 17 Chr
Cellulose
2.5 cm
22 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ Grade 17 Chr Cellulose Chromatography Paper > Type: sheet; L x W: 0.98 x 8.66 in. (2.5 x 22cm)