Learn More
Cytiva Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes
Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.
109.15 € - 388.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Bubble Point | 3.7bar |
Material | MCE |
Pore Size | 0.2 μm |
Flow Rate | 20s/100mL/12.5cm2 |
Product Code | Brand | Diameter (Metric) | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Sterility | Price | Quantity & Availability | ||||
11305324
|
Cytiva
10401731 |
142 mm | Non-sterile |
174.00 €
Pack of 25 |
In Stock
Log in to see stock. |
||||
11365314
|
Cytiva
10401712 |
47 mm | Non-sterile |
167.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
11325324
|
Cytiva
10401772 |
50 mm | Sterile |
168.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
11395314
|
Cytiva
10401726 |
110 mm | Non-sterile |
388.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
11375314
|
Cytiva
10401714 |
50 mm | Non-sterile |
165.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
11355314
|
Cytiva
10401706 |
25 mm | Non-sterile |
109.15 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
11385314
|
Cytiva
10401721 |
100 mm | Non-sterile |
348.00 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
Description
Enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. Whatman™ ME 24 Plain Mixed Cellulose Ester Membranes are composed of cellulose acetate and cellulose nitrate, which provides a more uniform surface.
- Supports visual enumeration of bacteria
- Appropriate for bacterial culturing, including E. coli culturing
- Biologically inert with good thermal stability
- Uniform microporous structure results in high flow rates when used for filtering
Specifications
Autoclavable | |
MCE | |
20s/100mL/12.5cm2 | |
Interleaving paper | |
135 μm |
3.7bar | |
0.2 μm | |
Plain | |
ME24 |
Safety and Handling
- ME24, 0.2 µm 142 mm, 25 pack
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.