missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes
Facilitates the counting process and minimizes eye fatigue with color contrast between the surface and the colonies. Whatman™ ME 25 Plain Mixed Cellulose Ester Membranes are characterized by a smooth and more uniform surface.
Brand: Cytiva 10401626
This item is not returnable.
View return policy
Description
- Excellent contrast for easier particle detection.
- Offers a high degree of internal surface area for greater adsorption of product.
- Higher dirt loading capacity.
- Biologically inert with good thermal stability.
- No surfactants to contaminate samples.
- Uniform microporous structure of membrane gives high flow rates.
Specifications
Autoclavable | |
Cellulose Acetate, Cellulose Nitrate | |
Non-sterile | |
Plain | |
ME25 | |
145 μm |
2.8bar | |
0.45 μm | |
12.5s/100mL/12.5cm2 | |
Interleaving paper | |
50Pack | |
110 mm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ME25, 0.45µm 110mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose