missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ MicroPlus MBS I Membrane Filtration Accessory: Funnel
Accessories for microbiological control filtration system
Brand: Cytiva 10445861
This item is not returnable.
View return policy
Description
- Autoclavable
- Presterilized
- 100mL
Specifications
Autoclavable | |
Microbiological filtration system I (MBS I) | |
100 mL |
Plastic funnel | |
Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction