missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Mixed Cellulose Ester Circle (ME28)

Brand:  Cytiva 10408472


Product Code. 11939217

  • 260.00 € / Pack of 100
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Mixed Cellulose Ester
1.2 μm
To quantitatively evaluate microorganism counts
Interleaving paper
0.31cm
266°F
Membrane Filter
1.96 in.
Green/Black
ME 28
Sterile
Hydrophilic
Medium
Grid
130°C
1.2 sec./100mL/12.5cm2
5 cm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 1
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers