missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes
Facilitates particle detection and minimizes eye fatigue with surface marks and color contrast. Whatman™ Mixed Cellulose Ester Membranes are available plain or gridded for automatic or manual counting procedures.
160.00 € - 228.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Mixed Cellulose Ester |
Includes | Interleaving paper |
Protein Binding Capacity | Medium |
Quantity | 100 Pack |
Product Code | Brand | Diameter (Metric) | Pore Size | Material (Membrane) | Waterflow Rate | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Material (Membrane) | Waterflow Rate | Price | Quantity & Availability | ||
11375284
|
Cytiva
10400912 |
47 mm | 0.8 μm | ME 27 | 2.8 sec./100mL/12.5cm2 |
166.00 €
Pack of 100 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
11315024
|
Cytiva
10400909 |
37 mm | 0.8 μm | ME 27 | 2.8 sec./100mL/12.5cm2 |
167.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||
11939207
|
Cytiva
10400772 |
50 mm | 3 μm | ME 29 | 1.2 s/100mL/12.5cm2 |
228.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||
11335014
|
Cytiva
10401512 |
47 mm | 0.6 μm | ME 26 | 4.8 sec./100mL/12.5cm2 |
160.00 €
Pack of 100 |
In Stock
Log in to see stock. |
||
Description
Offers a high degree of internal surface area for greater adsorption of product.
- Excellent contrast for easier particle detection.
- Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity.
- Black plain and black gridded membranes have a mix of cellulose nitrate and cellulose acetate.
- Higher dirt loading capacity.
- Biologically inert with good thermal stability.
- No surfactants to contaminate samples.
- Uniform microporous structure of membrane gives high flow rates.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
Specifications
Autoclavable | |
Interleaving paper | |
100 Pack | |
130°C |
Mixed Cellulose Ester | |
Medium | |
266°F | |
Membrane |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ME27, 0.8 µm 47 mm, 100 pack
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : Cellulose nitrate
- UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title