Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membranes - WME Range, 0.2 μm
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
Brand: Cytiva 7187-114
Description
Composed of cellulose acetate and cellulose nitrate membranes. Whatman™ Mixed Cellulose Ester Membranes - WME Range are characterized by a smoother and more uniform surface than pure nitrocellulose filters.
- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Color contrast provided by the filter surface also facilitates particle detection and minimizes eye fatigue.
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions

Specifications
Autoclavable | |
Mixed Cellulose Ester | |
0.2 μm | |
Sample preparation, microbiological studies, filtration of aqueous solutions | |
Pad | |
100 Pack | |
Grid | |
130°C | |
1.85 in. |
White | |
WME | |
Non-sterile | |
Black | |
Medium | |
3.1mm | |
266°F | |
White, Black Grid | |
47 mm |
Safety and Handling
- Danger
Hazard Category
- Flammable solid Category 1
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose
Your input is important to us. Please complete this form to provide feedback related to the content on this product.