missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Non-sterile ME 25 Mixed Cellulose Ester Membranes
Offers a larger adsorption of product with a high degree of internal surface area. Whatman™ Non-sterile ME25 Mixed Cellulose Membranes are available plain for automatic colony counting applications, as well as gridded to assist in manual counting procedures.
131.00 € - 167.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Mixed Cellulose Ester |
Material (Membrane) | ME 25 |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) | Grid Color | Size (Metric) Grid | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Grid Color | Size (Metric) Grid | Price | Quantity & Availability | |||
11365364
|
Cytiva
10406812 |
47 mm | Black | 3.1mm |
147.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
11325394
|
Cytiva
10409714 |
50 mm | White | 3.1mm |
167.00 €
Pack of 100 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
|||
11375364
|
Cytiva
10406814 |
50 mm | Black | 3.1mm |
151.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
10292131
|
Cytiva
10401612 |
47 mm | N/A | 3.1mm |
162.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
11365384
|
Cytiva
10409414 |
47 mm | Green/Black | 3.1mm |
134.00 €
Pack of 100 |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||
11325364
|
Cytiva
10406512 |
47 mm | Black | 5mm |
131.00 €
Pack of 100 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
Description
Offers a larger adsorption of product with a high degree of internal surface area. Whatman™ Non-sterile ME25 Mixed Cellulose Membranes are available plain for automatic colony counting applications, as well as gridded to assist in manual counting procedures.
Composed of cellulose acetate and cellulose nitrate characterized by a smooth and uniform surface. Color contrast provided by the filter surface facilitates particle detection and minimizes eye fatigue.- Gridded surface (3.1mm intervals) make filters ideal for colony counting after incubation
- Special nontoxic ink used is completely free from bacterial growth inhibitors
- Manufactured under strictly controlled cleanroom conditions
- Pore size: 0.45μm
Specifications
Autoclavable | |
ME 25 | |
Non-sterile | |
266°F | |
Membrane |
Mixed Cellulose Ester | |
0.45 μm | |
Medium | |
130°C | |
12.5 sec./100mL/12.5cm2 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ME25/21, 0.45µm 47mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose