Learn More
Cytiva Whatman™ Nuclepore™ Hydrophilic Membrane
A highly defined pore size cutoff in the sub-micron range enables particle deformity measurements, and the capture of extremely small particles
Brand:  Cytiva 10417001
56.67 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 16361396
  • 64.93 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Track-etched for highly defined pore size and high pore density
  • Treatment with polyvinylpyrrolidone (PVP) renders filters hydrophilic
  • Low extractables minimize filtration sample contamination risk
  • High chemical resistance and good thermal stability for a wide range of sample types
  • Low, consistent ash and low tare weights
  • Smooth flat membrane surface for high particle visibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Circles
Bioassays, Cell Biology and Environment Analysis
Polycarbonate Film Track-Etched (PFTE)
Track Etched Hydrophilic Membrane Filters
0.2 μm
Hydrophilic
100/Pk.
13 mm