missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Nytran™ Nylon Membranes

Cast uniformly on both sides of a support matrix, providing excellent symmetry and allowing membranes to lie flat without curling

Brand:  Cytiva 10416196

462.15 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10003401

  • 673.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Moderately charged—ideal for applications that require a lower charge including Southern and Northern blotting, colony and plaque lifts, as well as dot and slot blots
  • Allows for excellent signal-to-noise ratios
  • Binding capacity: >400μg/cm2
  • Compatible with isotopic and nonisotopic detection
  • Available in 0.2μm and 0.45μm pore size for optimal retention of oligos and larger DNA fragments

TRUSTED_SUSTAINABILITY
Specifications

Specifications

30 cm
Suitable for isotopic and nonisotopic detection
Roll
0.45 μm
3 m
>400μg/cm2
Nylon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.