Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
Marque: Cytiva 2612-990
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Spécification
1 to 12 | |
Routine pH testing | |
pH Indicator Test Strip, Integral Comparison Strip |
Strip | |
Plastic supports, paper segments on each strip, complementary indicator charts | |
100 x 11 mm |