Learn More
Cytiva Whatman™ pH Indicators Integral Comparison Strips
Used for rapid determination of pH values
Brand: Cytiva 2631-990
This item is not returnable.
View return policy
Description
Used for rapid determination of pH values
- Instant pH readings
- Accurate for wide range of routine pH testing
- Convenient and portable for field use
Specifications
9.5 to 12 | |
Routine pH testing | |
pH Indicator Test Strip, Integral Comparison |
Strip | |
Integral color chart | |
100 x 11 mm |