Learn More
Cytiva Whatman™ Polyamide Membranes
Universal filter made from pure polyamide for clarification and sterile filtration
Brand:  Cytiva 10414114
173.26 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 11358944
  • 216.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Mechanicall strong
  • Excellent wet/dry strength
  • Hydrophilic for aqueous and organic solutions
  • For up to 135°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyamide
Hydrophilic
50 mm
0.5 μm
110 μm