missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap™ Disposable Capsules - 36, 75, and 150 SPF
Provides serum prefiltration. Whatman™ Polycap™ Disposable Capsules permit filtration of tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspension. Includes inlet vent.
85.04 € - 901.00 €
Specifications
Autoclavable | Autoclavable |
---|---|
Material (Membrane) | Glass Microfiber, Polyethersulfone |
Pore Size | 1 μm |
Max. Pressure | 4.1 bar (60 psi) |
Product Code | Brand | Filtration Area | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filtration Area | Price | Quantity & Availability | |||||
11324614
|
Cytiva
6705-3600 |
260 cm2 |
85.04 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354614
|
Cytiva
6705-7500 |
535 cm2 |
104.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11379134
|
Cytiva
2820 |
1100 cm2 |
901.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Three-layer filter permits filtration of larger volumes
- Integrity testable
- With inlet vent
- Filter Media: Three layers: fine glass microfiber/ultrafine glass microfiber/1.0μm polysulfone
Serum Prefilter Capsules. Retention (with air purge), <2mL
For serum prefiltration. Handle tissue culture media, nutrients, immunologicals, enzymes, diagnostic standards/controls, biologicals, buffers, virus suspensions.
Specifications
Autoclavable | |
Glass Microfiber, Polyethersulfone | |
1 μm | |
4.1 bar (60 psi) |