missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Polycap Disposable Capsules, 75 AS
Polycap 75 AS filters for filtration of aqueous solutions
Brand: Cytiva 2707NS
This item is not returnable.
View return policy
Description
- Sterile and nonpyrogenic
- Integrity testable
- With inlet vent
- Filter Media: Low-extractable, hydrophilic nylon membrane and glass microfiber prefilter
- Filling bell model (09-744-61) minimizes contamination when filtering into bottle or flask
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris.
Specifications
Autoclavable | |
Nylon | |
Non-sterile | |
Polypropylene (PP) | |
4.1 bar (60 psi) |
Polycap AS 75 Capsule Filter, 0.45 μm, SB inlet and outlet (5 pcs) | |
0.45 μm | |
Filtration of aqueous solutions (such as cell culture media, serum enzymes and water) and removal of bacteria and debris. | |
Stepped Barb | |
Stepped Barb |