missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Polycap HD Disposable Capsules, 150 HD

Offers high filtration efficiency and excellent filtrate purity

Brand:  Cytiva 2813T

 View more versions of this product

Product Code. 11339114

  • 554.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Allows usage with broad range of solutions, pH, and temperature
  • High flow and high retention capacity
  • Able to be sterilized by autoclaving with steam at 121°C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polycap HD 150 Capsule Filter, 10.0 μm, with FNPT inlet and outlet (5 pcs)
Polycap HD 150
With Female NPT inlet/outlet
5/Pk.
10 μm
Polypropylene (PP)
4.1 bar (60 psi)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.