Learn More
Cytiva Whatman™ Polyester Drain Discs
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
Brand:  Cytiva 230500
 View more versions of this product
Product Code. 11919327
  • 73.92 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Provides a flat surface to eliminate filter tearing or rupturing. Whatman™ Polyester Drain Discs are used to support membrane filters. They are available in a variety of diameters for use in a range of devices.
For use with membrane hardware where extra support is needed for improved flow rate and throughput. Can be used as a separator between membranes in serial stack filtration applications.
  • Chemically inert polyester
  • 100μm Thickness
  • Binder-free
  • Available in several diameters
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyester
Membrane filtration, stack filtration
100 μm
22 mm