Learn More
Cytiva Whatman™ Potassium Iodide Test Paper on a Reel
Combines ease-of-use with unsurpassed accuracy and consistency
Brand:  Cytiva 2602-500A
Product Code. 10255831
  • 21.20 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
This rapid determination of pH values has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
For detecting chlorine and other oxidizing agents
Each
Potassium Iodide Test Paper
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • reel, lead acetate paper dispenser
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid, lead(2+) salt, trihydrate