missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.2μm BA83 Protran
100% pure nitrocellulose to ensure the highest binding capacity possible
159.00 € - 675.00 €
Specifications
For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
---|---|
Binding Capacity | High |
Material | Nitrocellulose |
Pore Size | 0.2 μm |
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Format | Quantity | Price | Quantity & Availability | |||
11305304
|
Cytiva
10401316 |
82mm dia. | Disc | 50/Pk. |
306.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
10589821
|
Cytiva
10401396 |
30cm x 3m | Roll | 1 Each | N/A | ||||
11345304
|
Cytiva
10401465 |
15 x 20cm | Sheet | 10/Pk. | N/A | ||||
11395324
|
Cytiva
10402452 |
20 x 20cm | Sheet | 5/Pk. | N/A | ||||
11905104
|
Cytiva
10402468 |
15cm x 3m | Roll | 1 Each | N/A | ||||
15530093
|
Cytiva
10401403 |
140 x 140mm | Sheet | 50/Pk. |
675.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
- Also available in circular formats
Specifications
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection | |
High | |
Nitrocellulose | |
0.2 μm |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- protran BA 83, 82mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose