missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Protran™ Nitrocellulose Blotting Membranes: 0.45μm BA85 Protran Circles
100% pure nitrocellulose to ensure the highest binding capacity possible
124.00 € - 995.00 €
Specifications
For Use With (Application) | Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection |
---|---|
Binding Capacity | High |
Format | Disc |
Material | Nitrocellulose |
Pore Size | 0.45 μm |
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Quantity | Price | Quantity & Availability | ||||
11305294
|
Cytiva
10401116 |
82 mm | 50/Pk. |
297.00 €
Pack of 50 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
11315294
|
Cytiva
10401124 |
132 mm | 25/Pk. | ||||||
11365334
|
Cytiva
10402506 |
25 mm | 100/Pk. |
124.00 €
Pack of 100 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
11385334
|
Cytiva
10402548 |
137 mm | 50/Pk. | ||||||
11375334
|
Cytiva
10402525 |
132 mm | 50/Pk. |
510.00 €
Pack of 50 |
Estimated Shipment: 03-10-2024
Log in to see stock. |
||||
11395334
|
Cytiva
10402578 |
25 mm | 1000/Pk. |
995.00 €
Pack of 1000 |
Estimated Shipment: 04-10-2024
Log in to see stock. |
||||
11305344
|
Cytiva
10402579 |
82.5 mm | 50/Pk. | ||||||
10087930
|
Cytiva
10405316 |
82 cm | 50/Pk. | ||||||
Description
- Universal membrane suitable for Western, Southern, and Northern blotting
- Hydrophilic, so no prewetting with methanol is required
- High binding capacity and very low background for superior signal-to-noise ratios without stringent washing conditions
- Compatible with all common detection methods, including radioactive, colorimetric, fluorescent, and chemiluminescent
- Provide excellent shelf-life for proteins—proteins maintain molecular recognition activity for up to five years on Protran membranes
- Three pore sizes available—0.45μm is ideal for larger molecular weight samples, 0.2μm ensures binding of samples below 20kDa, 0.1μm is used for molecules below 7kDa
Specifications
Suitable for radioactive, colorimetric, fluorescent, and chemiluminescent detection | |
Disc | |
0.45 μm |
High | |
Nitrocellulose |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- protran BA 85, 82mm
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose