Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 55
Consists of pure cellulose without any wetting agents.
Brand: Cytiva 10410224
This item is not returnable.
View return policy
Description
- Spontaneously wetting, very good wet strength
- Extremely chemically resistant; suitable for aqueous and organic media
- Hydrophilic
- Mechanically stable
- Sterilizable by all methods
Specifications
Regenerated Cellulose | |
Hydrophilic | |
110 mm |
0.5 μm | |
75 μm |