missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 58
Consists of pure cellulose without any wetting agents
Brand: Cytiva 10410314
242.12 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
Consists of pure cellulose without any wetting agents
- Spontaneously wetting, very good wet strength
- Extremely chemically resistant; suitable for aqueous and organic media
- Hydrophilic
- Mechanically stable
- Sterilizable by all methods
Specifications
Regenerated Cellulose | |
Hydrophilic | |
50 mm dia. |
0.2 μm | |
75 μm | |
50 mm |