Learn More
Cytiva Whatman™ Regenerated Cellulose Membranes: Grade RC 60
Consists of pure cellulose without any wetting agents
Brand:  Cytiva 10410014
308.75 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 11313365
  • 334.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Regenerated Cellulose
Hydrophilic
50 mm
1.0 μm
70 μm