missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached

For optimal performance in the laboratory. Wheaton™ 20mL PET Liquid Scintillation Vials, With Caps Attached offer low permeability to solvents and minimal background counts.

Brand:  DWK Life Sciences 986734

249.19 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12487351

  • 280.00 € / Pack of 500
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Clarity of glass with the safety of plastic
  • Vial can be incinerated; no harmful gas is generated
  • Vials offer low permeability to solvents and minimal background counts

 

Order Info

White. Packaged in utility trays with each partitioned tray holding 100 vials.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Linerless
White
PET
Offer low permeability to solvents and minimal background counts, vials can be safely incinerated, no harmful gas is generated
Tray
Polyethylene
61 mm
1.06 in.
27 mm
20 mL
2.4 in.
0.67 oz.
Screw Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers