missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 25mm Filtration Assemblies and Accessories

Choose from a wide range of filtration assemblies and accessories to suit your application needs

Brand:  DWK Life Sciences 419330

67.60 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12461571

  • 75.80 € / Each
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I, ASTM E 438 Type I, Class A
Glass
1 Case
15 mL
25mm filtration assembly
To handle small volumes of liquids for analysis of particulate or microbiological contamination
Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers