Learn More
DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert
For use with Wheaton CryoELITE™ Freestanding Cryogenic Vials
91.15 € - 389.00 €
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
12379959
|
DWK Life Sciences
W224621-0303 |
Sterile |
91.15 €
Pack of 100 |
Estimated Shipment: 19-08-2025
Log in to see stock. |
||||||
11561345
|
DWK Life Sciences
W985881 |
Non-sterile |
389.00 €
Pack of 500 |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
Description
- Makes sample identification and traceability reliable and convenient
- Bar code insert manually press fits into bottom of vial
- Data matrix readable with standard scanning hardware
- Made from same cryogenic radiation-grade, virgin polypropylene as vial, ensuring uniform thermal expansion
Specifications
Polypropylene | |
500/Cs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert