missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ 47mm Filtration Assembly and Accessories

Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.

Brand:  DWK Life Sciences 419370

384.75 EUR valid until 2025-12-31
Use promo code "24293" to get your promotional price.


 View more versions of this product

Product Code. 12411581

  • 405.00 € / Each
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed for filtering viscous or proteinaceous solutions to produce ultraclean filtrate. Wheaton™ 47mm Filtration Assembly and Accessories are manufactured from the highest quality materials.

  • Includes 300mL graduated funnel
  • Includes PTFE support screen gasket
  • Includes anodized clamp and a No. 8 silicone stopper
  • Includes a 316 stainless steel support with 120 mesh screen
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USP Type I, ASTM E 438 Type I
47mm filtration assembly
40 to 60 μm
One PTFE faced, coarse porosity (40 to 60μm) fritted glass support base, one PTFE faced 300mL graduated funnel, one anodized aluminum clamp, one No. 8 silicone stopper, hose connection for 0.25 in. I.D. tubing
Filtration Assembly, PTFE-faced Funnel
4.7 cm
0.25 in. hose
Borosilicate Glass
Autoclaving with the filter in place, analysis of particulate or microbiological contamination
1 Case
Up to 1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.