missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Fritted Glass Support Base for use with Wheaton 47mm Filtration Assembly
Replacement Glass Support Base with 40/35 [ST] Joint Connection for use with DWK Life Sciences Wheaton 47mm Glass Filter Holder Assembly (419380).
Brand: DWK Life Sciences 419382
332.18 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Fritted glass support base for Wheaton 47mm Vacuum Filter Holder Assemblies
- Manufactured from Wheaton 33 low extractable borosilicate glass which conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
- Coarse porosity (40 to 60μm)
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
Glass | |
Fritted Support Base |
47mm filtration assembly | |
Case of 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction