Learn More
DWK Life Sciences Wheaton™ Allihn Condensers For Soxhlet Extraction Apparatus
Used for continuous extraction of analytes from a solid into an organic solvent
Brand:  DWK Life Sciences 334003
212.80 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 12458942
  • 237.00 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Wetted parts are manufactured from WHEATON 33 low extractable borosilicate glass
  • How it works:
    • The flask containing the solvent is heated
    • Vapors rise in the larger outside tube
    • Vapors enter the water–cooled condenser and liquate
    • When the liquid level in the extractor reaches the top of the bent tube, siphoning action returns the extract-enriched solvent to the flask
    Compliance
    WHEATON 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
    TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    USP Type I and ASTM E438 Type I, Class A
    Soxhlet extraction apparatus
    34/45
    Allihn Condenser
    Used for continuous extraction of analytes from a solid into an organic solvent
    Larger outside tube and bent tube
    Borosilicate Glass