Learn More
DWK Life Sciences Wheaton™ Perforated Aluminum Capsules
GREENER_CHOICE
Product Code. 11718789 Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
13 mm
20 mm
Unit Size:
Pack of 1000
Product Code. 11718789
Brand: DWK Life Sciences 22419207
Only null left
Add to basket
Add to basket
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Tear-off seals remove completely allowing content to pour from bottle. Wheaton™ Tear-Off Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among many others
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Material Aluminum
Quantity 1000/Cs.
Type Complete Tear-off Cap
Closure Size 13 mm