missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with Metal Foil Liner

Product Code. 12403921 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
28-400
33-400
This item is not returnable. View return policy
This item is not returnable. View return policy

Features an aluminum foil faced pulp board liner that can be used with organic solvents

  • Good resistance to hydrocarbons, oils, ketones and alcohols
  • Not good for acids or alkalis
  • Closure size: 33mm-400
  • Phenolic
  • Autoclavable
  • Black

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 33-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Metal Foil
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.