missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner

Product Code. 10630375 Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
15-415
15-425
18-400
18-415
20-400
28-400
33-400
33-430
No. per Case:
100 Cs.
200 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

Features PTFE faced (14B) styrene-butadiene rubber liner

  • Designed for product safety
  • TFE/Styrene-Butadiene rubber liner
  • Screw cap
  • Pheolic
  • Black
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 33-430
Certifications/Compliance ISO 9001
Autoclavable Autoclavable
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.