Learn More
DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
Brand: DWK Life Sciences 240215
230.57 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.
- 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
- Aluminum foil faced pulp board liners can be used with organic solvents
- Use LDPE lined caps with distilled water, analytical standards and reagents
Specifications
18-400 | |
Screw Cap | |
Phenolic | |
Ideal for cell culture work |
Styrene Butadiene Rubber | |
Autoclavable | |
Black |