Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner
Provides completely inert inner seal and surface for sample or product
245.00 € - 489.00 €
Specifications
Product Type Screw Cap
Liner Material TFE, Styrene Butadiene Rubber
Autoclavable Autoclavable
Material Phenolic
Color Black
View More Specs
Products 2
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
11511235
View Documents
DWK Life Sciences
W240406
8-425
245.00 €
Pack of 200
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
12416171
View Documents
DWK Life Sciences
W242711
24-400
489.00 €
Pack of 200
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
Description
Description
  • Autoclavable
Specifications
Specifications
Screw Cap
Autoclavable
Black
TFE, Styrene Butadiene Rubber
Phenolic