missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ BOD Bottle without Stopper

Type I Borosilicate glass bottles are effective for a variety of applications

Brand:  DWK Life Sciences 227498

Product Code. 12493751

  • 414.00 € / Pack of 24
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Clear-glass allows easy viewing of bottle contents
  • Diameter: 69mm (2.72 in.)
  • Height: 143mm (5.63 in.)
  • Capacity: 300mL (10 oz.)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

No
Ideal for incubating diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during the stabilization of the decomposable organic matter by aerobic biochemical action
Borosilicate Glass
10.14 oz.
14.3 cm
6.9 cm
300 mL
Yes
Unnumbered
5.6 in.
2.7 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers