missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ BOD Bottles

Available with either glass robotic or pennyhead stoppers

Brand:  DWK Life Sciences 227497-01G

494.71 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11734189

  • 549.00 € / Pack of 24
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Available with either glass robotic or pennyhead stoppers

Bottles

  • Borosilicate glass, 60 or 300mL, numbered or unnumbered
  • With permanent marking areas; numbered versions are also bar-coded
  • Specially designed shoulder radius sweeps all air from inside the bottle during filling
  • 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
Stoppers
  • Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
  • Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
  • Stopper joint is compatible with the probes of the leading BOD and DO meters
  • Glass robotic stoppers are compatible with most robotic systems

Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.

Order Info

Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action
Borosilicate Glass
Pennyhead
6.49 in.
2.7 in.
300 mL
Yes
1 to 24
10.14 oz.
16.5 cm
6.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers