missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ BOD Bottles
Available with either glass robotic or pennyhead stoppers
Brand: DWK Life Sciences 227497-10G
494.71 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Available with either glass robotic or pennyhead stoppers
Bottles
- Borosilicate glass, 60 or 300mL, numbered or unnumbered
- With permanent marking areas; numbered versions are also bar-coded
- Specially designed shoulder radius sweeps all air from inside the bottle during filling
- 300mL bottles are recommended for the five-day BOD test referenced in Standard Methods for the Examination of Water and Wastewater, 18th Edition, 5210B, 1992
- Interchangeable borosilicate glass stoppers have tapered bottoms that also prevent air entrapment
- Flared mouth of bottle forms a water seal around stoppers that prevents air from being drawn in during incubation
- Stopper joint is compatible with the probes of the leading BOD and DO meters
- Glass robotic stoppers are compatible with most robotic systems
Use to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action.
Order Info
Dimensions: 300mL bottles are 2.75 diameter x 6.5 in. high (69 x 165mm); 60mL bottles are 1.75 diameter x 4.5 in. high (43 x 115mm)
Specifications
Yes | |
Used to incubate diluted samples of sewage, sewage effluents, polluted waters and industrial wastes to determine the amount of oxygen required during stabilization of decomposable organic matter by aerobic biochemical action | |
Borosilicate Glass | |
Pennyhead | |
6.49 in. | |
2.7 in. |
300 mL | |
Yes | |
217 to 240 | |
10.14 oz. | |
16.5 cm | |
6.9 cm |