missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Shop All Wheaton Science Products Products
Click to view available options
Material (Liner):
Linerless
Metal Foil
Polyethylene Disc
Polyseal™
Material:
Polyethylene
Polypropylene
Urea
Closure Size:
15-425
22-400
Unit Size:
Pack of 1000
Description
Screw Caps For Liquid Scintillation Vials
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
Specifications
Specifications
| Description | Cap Size: 22-400 |
| Material (Liner) | Polyethylene Disc |
| Color | White |
| Material | Urea |
| Type | Screw Cap |
| Closure Size | 22-400 |
| For Use With (Equipment) | Liquid Scintillation Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction