missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask

Product Code. p-4485893 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric) Cell:
5 mL
15 mL
Capacity (Metric) Media:
350 mL
1000 mL
For Use With (Application):
Adherent Cultures
Suspension Cultures
No. per Case:
1/Cs.
3/Cs.
5/Cs.
Unit Size:
Each
Pack of 3
Pack of 5
This item is not returnable. View return policy

Product Code. 12871793

Brand: DWK Life Sciences WCL10003

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

User manual with feeding and harvesting schedules

Membrane cell culture flasks ideal for antibody and protein production

High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.

  • Bulk media chamber allows for longer run times without media refreshing
  • Cell compartment concentrates cells, antibodies and proteins for simplified purification
  • For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.

02-912-745, 02-912-742

  • For adherent cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-745) or package three (02-912-742)
02-912-746, 02-912-743
  • For suspension cell cultivation
  • 350 mL media compartment
  • 5 mL cell compartment
  • Available as package of one (02-912-746) or package five (02-912-746)
02-912-744, 02-912-741
  • For suspension cell cultivation
  • 1000 mL media compartment
  • 15 mL cell compartment
  • Available as package of one (02-912-744) or package three (02-912-741)

TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) Cell 0.527 oz.
Capacity (English) Media 35.2 oz.
Capacity (Metric) Cell 15 mL
Capacity (Metric) Media 1000 mL
For Use With (Application) Suspension Cultures
Includes User manual with feeding and harvesting schedules
No. per Case 3/Cs.
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.