Learn More
DWK Life Sciences Wheaton™ Clear-Seal Connecting Adapters
Enable interfacing between standard semi-micro glassware and micro systems
Brand:  DWK Life Sciences 139479
48.29 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12425452
  • 49.35 € / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Feature a Clear-Seal™ inner joint on one end and a screw thread for the WHEATON Connection™ on the other end
  • 13-425 thread size comes with a screw cap and an O-ring to seal thermometers, gas inlet tubes, etc
  • Manufactured from WHEATON 33 low extractable borosilicate glass that conforms to ASTM E 438 Type I,Class A requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Screw cap, O-ring
Connecting Adapter
13-425
Micro systems
Borosilicate Glass
19/22
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Wheaton™ Clear-Seal Connecting Adapters > Connecting Adapters, Thermometer Adapter With O-ring, Cap: 13-425