missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
53-400 mm
58-400 mm
70-400 mm
89-400 mm
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Walls allow for complete removal of contents
- Wide-mouth design is great for large solid samples
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear USP Type III soda-lime glass
Specifications
Specifications
Capacity | 4 oz., 125 mL |
Capacity (Metric) | 125 mL |
Capacity (English) | 4 oz. |
Color | Clear |
Diameter (English) | 2.36 in. |
Material | Soda Lime Glass |
Closure Color | White |
Certifications/Compliance | USP Type III |
Closure Included | Yes |
Mouth | Wide |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction