missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps
Click to view available options
Capacity:
16 oz., 500 mL
2 oz., 60 mL
32 oz., 1000 mL
4 oz., 125 mL
8 oz., 250 mL
Diameter (English):
2.16 in.
2.36 in.
2.87 in.
3.58 in.
3.74 in.
Closure Size:
53-400 mm
58-400 mm
70-400 mm
89-400 mm
Height (English):
1.8 in.
2.6 in.
3.4 in.
3.8 in.
6.69 in.
Description
Ideal for soil sampling and environmental applications
Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass
- Walls allow for complete removal of contents
- Wide-mouth design is great for large solid samples
- Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage
Order Info
Available with caps attached or without caps. Caps can be ordered separately.
Compliance
Clear USP Type III soda-lime glass
Specifications
Specifications
Capacity | 2 oz., 60 mL |
Capacity (Metric) | 60 mL |
Capacity (English) | 2 oz. |
Color | Clear |
Diameter (English) | 2.16 in. |
Material | Soda Lime Glass |
Closure Color | White |
Height (Metric) | 48 mm |
Height (English) | 1.8 in. |
Closure Liner | PTFE |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'