Learn More
DWK Life Sciences Wheaton™ Complete Cyanide Distillation Apparatus: Components
Components for use with Wheaton Complete Cyanide Distillation Apparatus (cat. no. 09108)
Brand:  DWK Life Sciences 377164
 View more versions of this product
Product Code. 12481551
  • 163.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Compliance
For use according to ASTM D 2036
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyanide distillation kit
Cold Finger Jacket