missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits

Product Code. 17902631 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
OmniFlex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure
Ultra Pure Serum Stopper
Closure Color:
Blue
Red
Pakningsstørrelse:
1 set
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 17902631

Brand: DWK Life Sciences W991001CP

Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request

Certificeringer

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

TRUSTED_SUSTAINABILITY

Tekniske data

Closure Color Blue
Color Clear
Material Glass
Sterility Sterile
Volume (Metric) 2 mL
Includes Vial, Stopper, and Seal
No. per Case 278/Cs.
Product Line Wheaton CompletePAK
Volume (Metric) Total 2 mL
Closure Material Rubber (Stopper)
Closure Type OmniFlex 3G
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Type Sterile Kit
Closure Size 13 mm
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.